Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082032.1 | NP_001075501.1 | 7 - 8 | 1170 | 86 | |
XM_017347897.1 | XP_017203386.1 | 7 - 8 | 1108 | 86 | ||
GenBank mRNA | AF041202.1 | - | 7 - 8 | 1170 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Oryctolagus cuniculus Probe spans exons |