Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082044.1 | NP_001075513.1 | 29 - 30 | 3749 | 86 | |
XM_008259331.2 | XP_008257553.1 | 30 - 31 | 3763 | 86 | ||
XM_008259332.2 | XP_008257554.1 | 30 - 31 | 3762 | 86 | ||
XM_008259335.2 | XP_008257557.1 | 29 - 30 | 3723 | 86 | ||
XM_008259336.2 | XP_008257558.1 | 29 - 30 | 3724 | 86 | ||
XM_017343242.1 | XP_017198731.1 | 30 - 31 | 3765 | 86 | ||
GenBank mRNA | AF087468.1 | - | 29 - 30 | 3749 | 86 | |
AF417510.1 | - | 27 - 28 | 3476 | 86 | ||
AF417511.1 | - | 27 - 28 | 3476 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |