Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082150.2 | NP_001075619.1 | 2 - 3 | 756 | 83 | |
GenBank mRNA | AY191317.1 | - | 2 - 3 | 756 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |