Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082152.1 | NP_001075621.1 | 3 - 4 | 757 | 79 | |
XM_008263958.2 | XP_008262180.1 | 3 - 4 | 983 | 79 | ||
XM_008263959.2 | XP_008262181.1 | 2 - 3 | 416 | 79 | ||
GenBank mRNA | AY212510.1 | - | 3 - 4 | 757 | 79 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Oryctolagus cuniculus Probe spans exons |