Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082232.2 | NP_001075701.2 | 4 - 5 | 377 | 86 | |
XM_008272340.2 | XP_008270562.1 | 4 - 5 | 488 | 86 | ||
GenBank mRNA | AY829730.1 | - | 3 - 4 | 321 | 86 | |
J04712.1 | - | 3 - 4 | 348 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |