Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082296.1 | NP_001075765.1 | 4 - 5 | 248 | 146 | |
GenBank mRNA | M62387.1 | - | 4 - 5 | 248 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |