Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082302.2 | NP_001075771.1 | 270 | 97 | ||
GenBank mRNA | M64696.1 | - | 273 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Oryctolagus cuniculus |