Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082354.1 | NP_001075823.1 | 249 | 121 | ||
GenBank mRNA | U26725.2 | - | 249 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Oryctolagus cuniculus |