Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082381.1 | NP_001075850.1 | 1228 | 64 | ||
GenBank mRNA | U82163.1 | - | 1228 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus |