Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082711.1 | NP_001076180.1 | 3 - 4 | 334 | 69 | |
GenBank mRNA | AF026304.1 | - | 3 - 4 | 334 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Oryctolagus cuniculus Probe spans exons |