Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163072.1 | NP_001156544.1 | 2 - 3 | 227 | 69 | |
XM_017345505.1 | XP_017200994.1 | 2 - 3 | 215 | 69 | ||
GenBank mRNA | EU562212.1 | - | 2 - 3 | 227 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Probe spans exons |