Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL18R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164863.1 | NP_001158335.1 | 9 - 10 | 1268 | 104 | |
XM_008252716.2 | XP_008250938.1 | 10 - 11 | 1817 | 104 | ||
GenBank mRNA | EU731009.1 | - | 9 - 10 | 1268 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Oryctolagus cuniculus Probe spans exons |