Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160282.1 | NP_001153754.1 | 4 - 5 | 1039 | 70 | |
XM_008264005.2 | XP_008262227.1 | 5 - 6 | 1481 | 70 | ||
GenBank mRNA | FJ028656.1 | - | 4 - 5 | 1039 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |