Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712405.3 | XP_002712451.1 | 11 - 12 | 1465 | 100 | |
GenBank mRNA | AF307857.1 | - | 2 - 3 | 166 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Probe spans exons |