Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLA-DR-ALPHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171118.1 | NP_001164589.1 | 1 - 2 | 143 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |