Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171253.1 | NP_001164724.1 | 1 - 2 | 131 | 64 | |
XM_017344785.1 | XP_017200274.1 | 1 - 2 | 162 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Probe spans exons |