Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LENG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171392.1 | NP_001164863.1 | 137 | 65 | ||
XM_008251645.2 | XP_008249867.1 | 831 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus |