Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171419.1 | 372 | 81 | |||
XM_008274925.2 | XP_008273147.1 | 571 | 81 | |||
XM_008274926.2 | XP_008273148.1 | 572 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Oryctolagus cuniculus |