Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205047.1 | NP_001191976.1 | 6 - 7 | 983 | 88 | |
XM_008252739.2 | XP_008250961.1 | 6 - 7 | 1035 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |