Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266831.1 | NP_001253760.1 | 633 | 74 | ||
XM_015130398.2 | XP_014985884.1 | 821 | 74 | |||
XM_015130399.2 | XP_014985885.1 | 766 | 74 | |||
XM_028843118.1 | XP_028698951.1 | 662 | 74 | |||
XM_028843119.1 | XP_028698952.1 | 869 | 74 | |||
XM_028843120.1 | XP_028698953.1 | 723 | 74 | |||
XM_028843121.1 | XP_028698954.1 | 474 | 74 | |||
GenBank mRNA | JU320497.1 | - | 633 | 74 | ||
JU326026.1 | - | 523 | 74 | |||
JV637542.1 | - | 1576 | 74 | |||
JV703382.1 | - | 633 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |