Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194766.2 | NP_001181695.1 | 244 | 66 | ||
XM_015152920.1 | XP_015008406.1 | 244 | 66 | |||
GenBank mRNA | JU320504.1 | - | 178 | 66 | ||
JU474110.1 | - | 221 | 66 | |||
JV044838.1 | - | 249 | 66 | |||
JV601370.1 | - | 231 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |