Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047144.1 | NP_001040609.1 | 335 | 70 | ||
XM_015135921.2 | XP_014991407.2 | 427 | 70 | |||
XM_015135922.2 | XP_014991408.2 | 415 | 70 | |||
XM_015135923.2 | XP_014991409.2 | 462 | 70 | |||
GenBank mRNA | JU321545.1 | - | 426 | 70 | ||
JV635475.1 | - | 413 | 70 | |||
JV637049.1 | - | 355 | 70 | |||
JV669525.1 | - | 350 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |