Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040415.2 | NP_001035505.1 | 94 | 60 | ||
GenBank mRNA | AY913830.1 | - | 96 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |