Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260877.1 | NP_001247806.1 | 520 | 83 | ||
XM_015149293.2 | XP_015004779.1 | 664 | 83 | |||
XM_015149294.2 | XP_015004780.1 | 454 | 83 | |||
XM_015149295.2 | XP_015004781.1 | 383 | 83 | |||
GenBank mRNA | JU333058.1 | - | 639 | 83 | ||
JU333059.1 | - | 522 | 83 | |||
JU474397.1 | - | 550 | 83 | |||
JV046251.1 | - | 520 | 83 | |||
JV606622.1 | - | 544 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |