Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257802.1 | NP_001244731.1 | 345 | 118 | ||
XM_028853428.1 | XP_028709261.1 | 557 | 118 | |||
GenBank mRNA | JU322015.1 | - | 345 | 118 | ||
JV044564.1 | - | 342 | 118 | |||
JV490663.1 | - | 357 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |