Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260878.1 | NP_001247807.1 | 319 | 73 | ||
XM_028845799.1 | XP_028701632.1 | 4448 | 73 | |||
XM_028845802.1 | XP_028701635.1 | 375 | 73 | |||
GenBank mRNA | JU321836.1 | - | 327 | 73 | ||
JU470841.1 | - | 319 | 73 | |||
JV043412.1 | - | 300 | 73 | |||
JV601929.1 | - | 474 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |