Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKZF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258078.1 | NP_001245007.1 | 1093 | 62 | ||
XM_028830365.1 | XP_028686198.1 | 1157 | 62 | |||
XM_028830366.1 | XP_028686199.1 | 1230 | 62 | |||
XM_028830367.1 | XP_028686200.1 | 1785 | 62 | |||
XM_028830368.1 | XP_028686201.1 | 1267 | 62 | |||
XM_028830369.1 | XP_028686202.1 | 1821 | 62 | |||
XM_028830370.1 | XP_028686203.1 | 1163 | 62 | |||
XM_028830371.1 | XP_028686204.1 | 1079 | 62 | |||
XM_028830372.1 | XP_028686205.1 | 919 | 62 | |||
XM_028830373.1 | XP_028686206.1 | 953 | 62 | |||
GenBank mRNA | JU471109.1 | - | 1134 | 62 | ||
JU471110.1 | - | 1093 | 62 | |||
JU474708.1 | - | 1171 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |