Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLDIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261713.2 | NP_001248642.1 | 444 | 78 | ||
XM_015118842.2 | XP_014974328.1 | 482 | 78 | |||
GenBank mRNA | JU333919.1 | - | 411 | 78 | ||
JU475106.1 | - | 415 | 78 | |||
JV046712.1 | - | 444 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |