Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015150120.2 | XP_015005606.2 | 256 | 56 | ||
XM_015150129.2 | XP_015005615.2 | 691 | 56 | |||
XM_028847399.1 | XP_028703232.1 | 416 | 56 | |||
GenBank mRNA | JU336923.1 | - | 204 | 56 | ||
JU477655.1 | - | 202 | 56 | |||
JV048587.1 | - | 223 | 56 | |||
JV664505.1 | - | 224 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |