Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JCHAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260886.1 | NP_001247815.1 | 412 | 85 | ||
XM_015138542.2 | XP_014994028.1 | 552 | 85 | |||
XM_015138543.2 | XP_014994029.1 | 407 | 85 | |||
GenBank mRNA | JU475455.1 | - | 412 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |