Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257666.1 | NP_001244595.1 | 305 | 57 | ||
XM_015116858.2 | XP_014972344.1 | 359 | 57 | |||
GenBank mRNA | JU334744.1 | - | 305 | 57 | ||
JU475790.1 | - | 308 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |