Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260858.2 | NP_001247787.1 | 518 | 88 | ||
GenBank mRNA | JU321726.1 | - | 575 | 88 | ||
JU473527.1 | - | 685 | 88 | |||
JV043991.1 | - | 520 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |