Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU326875.1 | - | 684 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |