Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083346.3 | XP_001083346.2 | 2245 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |