Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194328.1 | NP_001181257.1 | 609 | 117 | ||
XM_015117397.2 | XP_014972883.1 | 763 | 117 | |||
GenBank mRNA | JU470696.1 | - | 592 | 117 | ||
JV636382.1 | - | 578 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |