Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PADI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265783.1 | NP_001252712.1 | 719 | 58 | ||
GenBank mRNA | JU332749.1 | - | 720 | 58 | ||
JV565421.1 | - | 740 | 58 | |||
JV667333.1 | - | 719 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |