Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193873.1 | NP_001180802.1 | 659 | 95 | ||
XM_015110974.2 | XP_014966460.2 | 490 | 95 | |||
XM_015110975.2 | XP_014966461.2 | 487 | 95 | |||
XM_015110976.1 | XP_014966462.1 | 651 | 95 | |||
XM_015110977.1 | XP_014966463.1 | 335 | 95 | |||
GenBank mRNA | JU335713.1 | - | 415 | 95 | ||
JU476597.1 | - | 413 | 95 | |||
JV047795.1 | - | 396 | 95 | |||
JV589402.1 | - | 390 | 95 | |||
JV600818.1 | - | 407 | 95 | |||
JV667440.1 | - | 403 | 95 |