Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258113.1 | NP_001245042.1 | 509 | 97 | ||
XM_015133842.2 | XP_014989328.1 | 1142 | 97 | |||
XM_015133843.2 | XP_014989329.1 | 1199 | 97 | |||
XM_015133844.2 | XP_014989330.1 | 9307 | 97 | |||
XM_015133845.2 | XP_014989331.1 | 862 | 97 | |||
XM_015133846.1 | XP_014989332.1 | 851 | 97 | |||
XM_015133847.1 | XP_014989333.1 | 641 | 97 | |||
XM_028844741.1 | XP_028700574.1 | 751 | 97 | |||
GenBank mRNA | JU324676.1 | - | 524 | 97 | ||
JU475328.1 | - | 509 | 97 | |||
JV046878.1 | - | 516 | 97 | |||
JV046879.1 | - | 516 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |