Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015153074.2 | XP_015008560.1 | 477 | 105 | ||
GenBank mRNA | JU321383.1 | - | 84 | 105 | ||
JV048306.1 | - | 73 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |