Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133580.2 | XP_014989066.2 | 1327 | 66 | ||
XM_028836972.1 | XP_028692805.1 | 1636 | 66 | |||
XM_028836997.1 | XP_028692830.1 | 1434 | 66 | |||
GenBank mRNA | JU326270.1 | - | 1111 | 66 | ||
JU326271.1 | - | 1111 | 66 | |||
JV634422.1 | - | 1129 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |