Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015121306.2 | XP_014976792.2 | 1009 | 89 | ||
XM_015121307.2 | XP_014976793.2 | 1531 | 89 | |||
XM_028837208.1 | XP_028693041.1 | 945 | 89 | |||
XM_028837209.1 | XP_028693042.1 | 845 | 89 | |||
XM_028837210.1 | XP_028693043.1 | 809 | 89 | |||
GenBank mRNA | JV704846.1 | - | 914 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |