Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265846.1 | NP_001252775.1 | 200 | 67 | ||
GenBank mRNA | JU334324.1 | - | 200 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |