Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLGN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278449.1 | NP_001265378.1 | 306 | 73 | ||
XM_015118450.2 | XP_014973936.1 | 1198 | 73 | |||
XM_028835458.1 | XP_028691291.1 | 1120 | 73 | |||
XM_028835459.1 | XP_028691292.1 | 864 | 73 | |||
GenBank mRNA | AF462607.1 | - | 44 | 73 | ||
JV552452.1 | - | 145 | 73 | |||
JV623614.1 | - | 167 | 73 | |||
JV691211.1 | - | 230 | 73 | |||
JV719711.1 | - | 165 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |