Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032865.1 | NP_001028037.1 | 791 | 67 | ||
GenBank mRNA | AF240771.1 | - | 791 | 67 | ||
JV518189.1 | - | 838 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |