Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037282.2 | NP_001032359.1 | 319 | 95 | ||
NM_001110260.1 | NP_001103730.1 | 319 | 95 | |||
XM_028852101.1 | XP_028707934.1 | 786 | 95 | |||
GenBank mRNA | AF466351.1 | - | 319 | 95 | ||
AF466352.1 | - | 319 | 95 | |||
AY513587.1 | - | 107 | 95 | |||
AY528238.1 | - | 293 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |