Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195397.3 | NP_001182326.2 | 614 | 125 | ||
GenBank mRNA | AJ000067.1 | - | 462 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |