Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038659.1 | NP_001033748.1 | 164 | 111 | ||
GenBank mRNA | DQ191051.1 | - | 164 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |