Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257684.1 | NP_001244613.1 | 382 | 92 | ||
GenBank mRNA | JU332614.1 | - | 382 | 92 | ||
JV604024.1 | - | 402 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |