Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260801.1 | NP_001247730.1 | 733 | 141 | ||
XM_015134811.2 | XP_014990297.1 | 794 | 141 | |||
GenBank mRNA | JU320007.1 | - | 733 | 141 | ||
JU320008.1 | - | 733 | 141 | |||
JV662501.1 | - | 758 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |