Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266760.1 | NP_001253689.1 | 460 | 129 | ||
XM_015144113.2 | XP_014999599.1 | 439 | 129 | |||
XM_028850263.1 | XP_028706096.1 | 495 | 129 | |||
XM_028850264.1 | XP_028706097.1 | 438 | 129 | |||
GenBank mRNA | JU320610.1 | - | 442 | 129 | ||
JU474016.1 | - | 433 | 129 | |||
JV045946.1 | - | 460 | 129 | |||
JV640371.1 | - | 432 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |